Anti-HSD17B2 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-3856R-CY5
10395-942EA 811.1 USD
10395-942
Anti-HSD17B2 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Capable of catalyzing the interconversion of testosterone and androstenedione, as well as estradiol and estrone. Also has 20-alpha-HSD activity. Uses NADH while EDH17B3 uses NADPH.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HSD17B2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR