Anti-TNFRSF19 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-3852R-A647
10388-768EA 811.1 USD
10388-768
Anti-TNFRSF19 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
TROY is a member of the TNF receptor superfamily. This receptor is highly expressed during embryonic development. It has been shown to interact with TRAF family members, and to activate the JNK signaling pathway when overexpressed in cells. This receptor is capable of inducing apoptosis by a caspase-independent mechanism, and it is thought to play an essential role in embryonic development. Alternatively spliced transcript variants encoding distinct isoforms have been described.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TNFRSF19
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR