Anti-TNFSF9 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-3851R-A647
10388-744EA 776.17 USD
10388-744
Anti-TNFSF9 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Cytokine that binds to TNFRSF9. Induces the proliferation of activated peripheral blood T-cells. May have a role in activation-induced cell death (AICD). May play a role in cognate interactions between T-cells and B-cells/macrophages.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TNFSF9
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR