Anti-EIF3H Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-3842R-CY5
10388-614EA 776.17 USD
10388-614
Anti-EIF3H Rabbit Polyclonal Antibody (Cy5®)
Antibodies
EIF3S3 binds to the 40S ribosome and promotes the binding of methionyl-tRNAi and mRNA. It associates with the p170 subunit of EIF3. The EIF3 is composed of at least 12 different subunits.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EIF3H
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR