Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EIF3H Rabbit Polyclonal Antibody (Biotin)

Supplier: Bioss

BS-3842R-BIO
10388-610EA 504.09 USD InStock
10388-610
Anti-EIF3H Rabbit Polyclonal Antibody (Biotin)
Antibodies
EIF3S3 binds to the 40S ribosome and promotes the binding of methionyl-tRNAi and mRNA. It associates with the p170 subunit of EIF3. The EIF3 is composed of at least 12 different subunits.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: EIF3H
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty