Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TSC1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-3837R-CY5
10388-550
Anti-TSC1 Rabbit Polyclonal Antibody (Cy5®)
Hamartin, or TSC1, is a suspected tumor suppressor implicated in the disease tuberous sclerosis 1. It is a negative regulator of cell division controlling the transition from G0/G1 to S phase, and it seems to act through the phosphatidylinositol 3 kinase/Akt pathway. TSC1 interacts with tuberin m(TSC2), which is thought to be a GAP (GTPase Activating Protein) for the Rap1 and Rab5 small G Proteins. The Hamartin/Tuberin complex has been shown to inhibit mTor. Hamartin has also been shown to interact with ERM (Ezrin-Radixin-Moesin) proteins and with F-actin, suggesting a role for TSC proteins in modulation of cell adhesion and morphology.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TSC1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty