Anti-TSC1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-3837R-A555
10388-542EA 746.42 USD
10388-542
Anti-TSC1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Hamartin, or TSC1, is a suspected tumor suppressor implicated in the disease tuberous sclerosis 1. It is a negative regulator of cell division controlling the transition from G0/G1 to S phase, and it seems to act through the phosphatidylinositol 3 kinase/Akt pathway. TSC1 interacts with tuberin m(TSC2), which is thought to be a GAP (GTPase Activating Protein) for the Rap1 and Rab5 small G Proteins. The Hamartin/Tuberin complex has been shown to inhibit mTor. Hamartin has also been shown to interact with ERM (Ezrin-Radixin-Moesin) proteins and with F-actin, suggesting a role for TSC proteins in modulation of cell adhesion and morphology.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TSC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR