Anti-MRPL12 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-3834R-A488
10388-468EA 811.1 USD
10388-468
Anti-MRPL12 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
MRPL12 is a component of the 39S subunit of mammalian mitochondrial ribosomes. This protein forms homodimers whereas in prokaryotic ribosomes two L7/L12 dimers and one L10 protein form the L8 protein complex.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MRPL12
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR