Anti-RHEB Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-3832R-CY5.5
10388-456EA 811.1 USD
10388-456
Anti-RHEB Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Stimulates the phosphorylation of S6K1 and EIF4EBP1 through activation of mTORC1 signaling. Activates the protein kinase activity of mTORC1. Has low intrinsic GTPase activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RHEB
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR