Anti-PNPLA2 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-3831R-CY5
10388-430EA 810.85 USD
10388-430
Anti-PNPLA2 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Catalyzes the initial step in triglyceride hydrolysis in adipocyte and non-adipocyte lipid droplets. Also has acylglycerol transacylase activity. May act coordinately with LIPE/HLS within the lipolytic cascade. Regulates adiposome size and may be involved in the degradation of adiposomes. May play an important role in energy homeostasis. May play a role in the response of the organism to starvation, enhancing hydrolysis of triglycerides and providing free fatty acids to other tissues to be oxidized in situations of energy depletion.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PNPLA2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR