Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SDH Rabbit Polyclonal Antibody (Biotin)

Supplier: Bioss

BS-3829R-BIO
10394-008
Anti-SDH Rabbit Polyclonal Antibody (Biotin)
Sorbitol dehydrogenase (SDH), a member of the medium-chain dehydrogenase/reductase protein family and the second enzyme of the polyol pathway of glucose metabolism, converts sorbitol to fructose strictly using NAD(+) as coenzyme. SDH is expressed almost ubiquitously in all mammalian tissues. The enzyme has attracted considerable interest due to its implication in the development of diabetic complications as the polyol pathway is particularly active in hyperglycemic states. Although SORD is closely related to the class I long-chain alcohol dehydrogenases, it differs in substrate specificity, catalyzing polyols such as sorbitol and xylitol but having no activity towards primary alcohols.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: SDH
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty