Anti-HNF4A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-3828R-FITC
10393-994EA 811.1 USD
10393-994
Anti-HNF4A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Transcriptionally controlled transcription factor. Binds to DNA sites required for the transcription of alpha 1-antitrypsin, apolipoprotein CIII, transthyretin genes and HNF1-alpha. May be essential for development of the liver, kidney and intestine.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HNF4A
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR