Anti-GDF15 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-3818R-CY5
10388-254EA 811.1 USD
10388-254
Anti-GDF15 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Growth differentiation factor 15 (GDF15) is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. It has been implicated in a variety of functions directly related to tumorigenicity including antiproliferative and pro-apoptotic effects. BMP proteins are secreted growth factors that are characterized by seven conserved cysteine residues. In general, they are regulators of cell growth and differentiation in both embryonic and adult tissues.GDF15 is an important downstream mediator of DNA damage signaling and a transcriptional target of p53.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GDF15/MIC-1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR