Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NEDD8 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-3812R-A647
10388-152EA 580.76 USD InStock
10388-152
Anti-NEDD8 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Ubiquitin like proteins fall into two classes: the first class, ubiquitin-like modifiers (UBLs) function as modifiers in a manner analogous to that of ubiquitin. Examples of UBLs are SUMO, Rub1 (also called Nedd8), Apg12 and Hub1. Proteins of the second class include Parkin, RAD23 and DSK2, are designated ubiquitin domain proteins (UDPs). These proteins contain domains that are related to ubiquitin but are otherwise unrelated to each other. In contrast to UBLs, UDPs are not proteolytically processed or conjugated to other proteins. Rub1 and the corresponding human homolog NEDD8 are activated by the E1 ubiquitin activating enzyme UBA2, that forms isopeptide linkages between thio esters. NEDD8 shows 80% homology to ubiquitin. The best known targets of Rub1 modification are members of the cullin family. Cullins are subunits of an E3 ubiquitin ligase complex called the Skp1 /Cul1/Cdc53 F box (SCF). The SCF promotes transfer of ubiquitin from a ubiquitin conjugating enzyme (E2) to the target protein. Rub1 modification may regulate SCF function or localization.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NEDD8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty