Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PLAA Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-3807R-CY5.5
10388-104
Anti-PLAA Rabbit Polyclonal Antibody (Cy5.5®)
Phospholipase A2 activator protein (PLAP), activates PLA2 (phospholipase A2 enzyme) and is an important mediator of eicosanoid generation. Also known as a pro-inflammatory agent, PLAP has been found in inflamed tissues and synovial fluid from patients with rheumatoid arthritis. As such, it is believed to play an important role in the regulation of inflammatory diseases. The formation of PLAP can be stimulated by IL-1 beta and TNF-alpha.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PLAA
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty