Anti-PLAA Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-3807R-A555
10388-094EA 811.1 USD
10388-094
Anti-PLAA Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Phospholipase A2 activator protein (PLAP), activates PLA2 (phospholipase A2 enzyme) and is an important mediator of eicosanoid generation. Also known as a pro-inflammatory agent, PLAP has been found in inflamed tissues and synovial fluid from patients with rheumatoid arthritis. As such, it is believed to play an important role in the regulation of inflammatory diseases. The formation of PLAP can be stimulated by IL-1 beta and TNF-alpha.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PLAA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR