Anti-UCHL1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-3806R-A350
10388-066EA 811.1 USD
10388-066
Anti-UCHL1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Ubiquitin-protein hydrolase involved both in the processing of ubiquitin precursors and of ubiquitinated proteins. This enzyme is a thiol protease that recognizes and hydrolyzes a peptide bond at the C-terminal glycine of ubiquitin. Also binds to free monoubiquitin and may prevent its degradation in lysosomes. The homodimer may have ATP-independent ubiquitin ligase activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: UCHL1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR