Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MB Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-3805R-CY3
10388-052EA 596.85 USD InStock
10388-052
Anti-MB Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Serves as a reserve supply of oxygen and facilitates the movement of oxygen within muscles.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MB
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty