Anti-ZNF772 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-3803R-A488
10388-004EA 811.1 USD
10388-004
Anti-ZNF772 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Mammaglobin B may bind androgens and other steroids, it may also bind estramustine, a chemotherapeutic agent used for prostate cancer. It may be under transcriptional regulation of steroid hormones.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZNF772
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR