Anti-IgL Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-3801R-A488
10393-898EA 775.93 USD
10393-898
Anti-IgL Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
All five immunoglobulin classes share the same basic four polypeptide chain structure of two heavy-chains and two light chains. There are five heavy chain types, and two light-chain types (Kappa and Lambda) both having a molecular weight of 22.5kDa. Any heavy-chain type can associate with either light-chain type, but on any immunoglobulin molecule both light-chains are of the same type. Kappa and Lambda consist of a variable region and a constant region and can easily be differentiated by the antigenic properties of the constant region.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IgL
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope: Light Chain
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR