Anti-CRX Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-3798R-CY7
10387-978EA 811.1 USD
10387-978
Anti-CRX Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Transcription factor that binds and transactivates the sequence 5'-TAATC[CA]-3' which is found upstream of several photoreceptor-specific genes, including the opsin genes. Acts synergistically with other transcription factors, such as NRL, RORB and RAX, to regulate photoreceptor cell-specific gene transcription. Essential for the maintenance of mammalian photoreceptors.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRX
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR