Anti-CRX Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-3798R-A488
10387-964EA 776.17 USD
10387-964
Anti-CRX Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Transcription factor that binds and transactivates the sequence 5'-TAATC[CA]-3' which is found upstream of several photoreceptor-specific genes, including the opsin genes. Acts synergistically with other transcription factors, such as NRL, RORB and RAX, to regulate photoreceptor cell-specific gene transcription. Essential for the maintenance of mammalian photoreceptors.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRX
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR