Anti-surface layer protein Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-3797R-FITC
10387-956EA 811.1 USD
10387-956
Anti-surface layer protein Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
surface layer protein

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: surface layer protein
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR