Anti-surface layer protein Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-3797R-CY3
10387-948EA 811.1 USD
10387-948
Anti-surface layer protein Rabbit Polyclonal Antibody (Cy3®)
Antibodies
surface layer protein

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: surface layer protein
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR