Anti-ANO1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-3794R-CY5
10394-900EA 811.1 USD
10394-900
Anti-ANO1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Calcium-activated chloride channel (CaCC) which plays a role in transepithelial anion transport and smooth muscle contraction. Required for the normal functioning of the interstitial cells of Cajal (ICCs) which generate electrical pacemaker activity in gastrointestinal smooth muscles. Acts as a major contributor to basal and stimulated chloride conductance in airway epithelial cells and plays an important role in tracheal cartilage development.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ANO1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR