Anti-PERIA Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-3789R-CY3
10387-866EA 776.17 USD
10387-866
Anti-PERIA Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Modulator of adipocyte lipid metabolism. Coats lipid storage droplets to protect them from breakdown by hormone-sensitive lipase (HSL). Its absence may result in leanness. Plays a role in unilocular lipid droplet formation by activating CIDEC. Their interaction promotes lipid droplet enlargement and directional net neutral lipid transfer. May modulate lipolysis and triglyceride levels.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PERIA
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR