Anti-STK17B Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-3785R-CY7
10387-776EA 811.1 USD
10387-776
Anti-STK17B Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Apoptosis is mediated by death domain containing adapter molecules and a caspase family of proteases. Certain serine/threonine protein kinases, such as ASK1 and RIP, are mediators of apoptosis. Two novel serine/threonine kinases that induce apoptosis were recently identified and designated DRAK1 and DRAK2 (for DAP kinase related apoptosis inducing protein kinases). DRAKs contain an N terminal kinase domain and a C terminal regulation domain. Overexpression of DRAK2 induces apoptosis. DRAKs have high sequence homology to DAP and ZIP kinases, and they represent a novel family of serine/threonine kinases, which mediates apoptosis through their catalytic activities. DRAK2 is located in nucleus and the messenger RNA was ubiquitously expressed in human tissues.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: STK17B
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR