Anti-ASF1A Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-3777R-CY5
10387-668EA 775.93 USD
10387-668
Anti-ASF1A Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Histone chaperone that facilitates histone deposition and histone exchange and removal during nucleosome assembly and disassembly. Cooperates with chromatin assembly factor 1 (CAF-1) to promote replication-dependent chromatin assembly and with HIRA to promote replication-independent chromatin assembly. Required for the formation of senescence-associated heterochromatin foci (SAHF) and efficient senescence-associated cell cycle exit.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ASF1A
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR