Anti-S100 A4 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-3759R-CY5
10394-788EA 776.17 USD
10394-788
Anti-S100 A4 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes. S100A4 has been implicated in motility, invasion and tubulin polymerization. It is also involved in tumor metastasis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: S100 A4
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR