Anti-PP1A/PP2CA/PPP1CA Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-3756R-A647
10387-382EA 810.85 USD
10387-382
Anti-PP1A/PP2CA/PPP1CA Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
PPP1A is a serine/threonine protein phosphatase catalytic subunit that is essential for regulating cellular stress responses in eukaryotes. It binds to magnesium or manganese ions and exists as a monomer. It is essential for cell division, and participates in the regulation of glycogen metabolism, muscle contractility and protein synthesis. PPP1A is involved in the regulation long term synaptic plasticity and may play an important role in dephosphorylating substrates such as Ca2+/calmodulin dependent protein kinase II.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PP1A/PP2CA/PPP1CA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR