Anti-CRKL Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-3755R-A350
10387-352EA 776.17 USD
10387-352
Anti-CRKL Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Crkl is a 34kDa adaptor protein which has been shown to activate the RAS and JUN kinase signalling pathways and transforms fibroblasts in a RAS dependant manner. Crkl is a substitute of BCR ABL tyrosine kinase. In addition, Crkl has oncogenic potential.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRKL
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR