Anti-CLDN6 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-3754R
10387-326EA 649.69 USD
10387-326
Anti-CLDN6 Rabbit Polyclonal Antibody
Antibodies
Claudin 6 is a multipass transmembrane protein in the Claudin family. Claudin 6 is expressed by epithelial cells where it participates in tissue development and the maintenance of tight junction integrity. Claudin 6 is also a global marker of definitive endoderm. Directly interacts with TJP1/ZO-1, TJP2/ZO-2 and TJP3/ZO-3.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CLDN6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR