Anti-STMN2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-3753R-A350
10387-304EA 746.42 USD
10387-304
Anti-STMN2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Regulator of microtubule stability. When phosphorylated by MAPK8, stabilizes microtubules and consequently controls neurite length in cortical neurons. In the developing brain, negatively regulates the rate of exit from multipolar stage and retards radial migration from the ventricular zone (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: STMN2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR