Anti-RAN Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-3751R-HRP
10394-748EA 776.17 USD
10394-748
Anti-RAN Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Ran (ras-related nuclear protein) is a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The Ran protein is also involved in control of DNA synthesis and cell cycle progression. Nuclear localization of Ran requires the presence of regulator of chromosome condensation 1 (RCC1). Mutations in Ran disrupt DNA synthesis. Because of its many functions, it is likely that Ran interacts with several other proteins. Ran regulates formation and organization of the microtubule network independently of its role in the nucleus-cytosol exchange of macromolecules. Ran could be a key signaling molecule regulating microtubule polymerization during mitosis. RCC1 generates a high local concentration of Ran-GTP around chromatin which, in turn, induces the local nucleation of microtubules. Ran is an androgen receptor (AR) coactivator that binds differentially with different lengths of polyglutamine within the androgen receptor. Polyglutamine repeat expansion in the AR is linked to Kennedy's disease (X-linked spinal and bulbar muscular atrophy). Ran coactivation of the AR diminishes with polyglutamine expansion within the AR, and this weak coactivation may lead to partial androgen insensitivity during the development of Kennedy's disease.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: RAN
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR