Anti-PNPLA1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-3742R-A350
10387-144EA 640.91 USD
10387-144
Anti-PNPLA1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
PNPLA1 functions as a lipid hydrolase. Human patatin-like phospholipases, such as PNPLA1, have been implicated in regulation of adipocyte differentiation and have been induced by metabolic stimuli.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PNPLA1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR