Anti-HSF1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-3741R-A488
10393-760EA 776.17 USD
10393-760
Anti-HSF1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
DNA-binding protein that specifically binds heat shock promoter elements (HSE) and activates transcription. In higher eukaryotes, HSF is unable to bind to the HSE unless the cells are heat shocked.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HSF1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR