Anti-CLDN6 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-3736R-CY3
10387-082EA 811.12 USD
10387-082
Anti-CLDN6 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Claudin 6 is a multipass transmembrane protein in the Claudin family. Claudin 6 is expressed by epithelial cells where it participates in tissue development and the maintenance of tight junction integrity. Claudin 6 is also a global marker of definitive endoderm. Directly interacts with TJP1/ZO-1, TJP2/ZO-2 and TJP3/ZO-3.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CLDN6
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR