Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PRKDC Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-3735R-CY5
10387-060
Anti-PRKDC Rabbit Polyclonal Antibody (Cy5®)
DNA PK is the key component of the non-homologous end-joining (NHEJ) pathway of DSB repair in mammalian cells. DNA PK consists of a heterodimeric DNA-binding subunit (Ku70/80) and an approximately 465 kDa catalytic subunit (DNA PKcs). DNA PKcs is a serine/threonine protein kinase whose activity is greatly stimulated by its recruitment to DNA breaks by the Ku heterodimer.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PRKDC
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty