Anti-Talin Rabbit Polyclonal Antibody (Biotin)

BS-3719R-BIO
10396-140EA 559.62 USD
10396-140
Anti-Talin Rabbit Polyclonal Antibody (Biotin)
Probably involved in connections of major cytoskeletal structures to the plasma membrane. High molecular weight cytoskeletal protein concentrated at regions of cell-substratum contact and, in lymphocytes, at cell-cell contacts (By similarity).

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: Talin
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR