Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TP73 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-3717R-A488
10386-754EA 611.42 USD InStock
10386-754
Anti-TP73 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
P73 protein is a structural and functional homologue of p53, a tumor suppressor gene. In this study, The p73 protein, p19ras, by the yeast two-hybrid screening method. Alternative splicing of the proto-oncogene H-ras pre-mRNA has led to two distinct transcripts, Ras proteins are known to be small membrane-localized guanine nucleotide-binding proteins. However, unlike other Ras proteins, p19ras is localized in the nucleus and the cytosol and its interaction with P73 protein occurred exclusively in the nucleus. Oncogenic MDM2 (mouse double minutes 2) is a known repressor of p73 transcriptional activity. In this study, when p19ras was bound to MDM2, it further inhibited the association of MDM2 to the p73 protein. Therefore, this study presents a novel pathway of Ras signaling that occurs in the nucleus, involving p19ras and p73.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TP73
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty