Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CTR9 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-3697R-FITC
10386-400
Anti-CTR9 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
CTR9 is a component of the PAF (polymerase-associated factor) complex which interacts with histone-modifying enzymes and RNA polymerase II and plays a role in a number of transcription-related processes. Recent studies show that CTR9 can regulate IL-6-responsive gene transcription by influencing histone methylation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CTR9
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty