Anti-CTR9 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-3697R-CY5.5
10386-396EA 811.1 USD
10386-396
Anti-CTR9 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
CTR9 is a component of the PAF (polymerase-associated factor) complex which interacts with histone-modifying enzymes and RNA polymerase II and plays a role in a number of transcription-related processes. Recent studies show that CTR9 can regulate IL-6-responsive gene transcription by influencing histone methylation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CTR9
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR