Anti-KPNB1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-3695R-A350
10386-326EA 776.17 USD
10386-326
Anti-KPNB1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
NTF97 is involved in nuclear protein import, either by associating itself with an adapter protein (for example, importin-alpha subunit which binds to nuclear localization signals (NLS) in cargo substrates), or by acting autonomously as a nuclear transport receptor (serves as NLS receptor, docking of the importin/substrate complex to the nuclear pore complex).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KPNB1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR