Anti-DIO2 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-3673R-CY5.5
10373-904EA 810.85 USD
10373-904
Anti-DIO2 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
DIO2 belongs to the iodothyronine deiodinase family and is responsible for the deiodination of T4 (3,5,3',5'-tetraiodothyronine) into T3 (3,5,3'-triiodothyronine). It is essential for providing the brain with appropriate levels of T3 during the critical period of development. DIO2 is expressed in heart, skeletal muscle, placenta, fetal brain and several regions of the adult brain. There are two named isoforms.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DIO2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR