Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSPB8 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-3672R-CY3
10371-708EA 580.76 USD InStock
10371-708
Anti-HSPB8 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Hsp22 (HSPB8) is a 196-amino acid protein that contains a central portion homologous to a highly conserved HSP-alpha crystallin domain common to all the small heat shock protein (HSP20) family members. Hsp22 is a monomeric protein which interacts with HSPB1. It displays temperature-dependent chaperone activity. The highest abundance of Hsp22 is in skeletal muscle, heart, and placenta. Mutations in the HSPB8 gene are associated with the inherited peripheral neuropathies, autosomal dominant distal hereditary motor neuropathy type IIA (dSMA) and axonal Charcot-Marie-Tooth disease type 2L (CMT2L).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HSPB8
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty