Anti-SLC44A1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-3667R-CY7
10371-600EA 811.1 USD
10371-600
Anti-SLC44A1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Choline transporter. May be involved in membrane synthesis and myelin production.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC44A1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR