Anti-CASP7 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-3665R-A488
10371-546EA 811.1 USD
10371-546
Anti-CASP7 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
This gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. The precursor of this caspase is cleaved by caspase3 and 10. It is activated upon cell death stimuli and induces apoptosis. Alternative splicing results in four transcript variants, encoding three distinct isoforms. [provided by RefSeq].The antibody should recognize both pro-form and p20 cleaved-form. The antibody does not cross-react with other Caspase family members.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CASP7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR