Anti-WNT10B Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-3662R-CY5
10375-970EA 811.1 USD
10375-970
Anti-WNT10B Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: WNT10B
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR