Anti-DACH2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-3659R-HRP
10375-930EA 776.17 USD
10375-930
Anti-DACH2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
DACH2 is a transcription factor that is involved in regulation of organogenesis. It seems to be a regulator for SIX1 and SIX6 and may act as a corepressor of SIX6 in regulating proliferation by directly repressing cyclin-dependent kinase inhibitors, including the p27Kip1 promoter. It probably binds to DNA via its DACH box-N domain.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: DACH2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR