Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DACH2 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-3659R-CY7
10375-926
Anti-DACH2 Rabbit Polyclonal Antibody (Cy7®)
DACH2 is a transcription factor that is involved in regulation of organogenesis. It seems to be a regulator for SIX1 and SIX6 and may act as a corepressor of SIX6 in regulating proliferation by directly repressing cyclin-dependent kinase inhibitors, including the p27Kip1 promoter. It probably binds to DNA via its DACH box-N domain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DACH2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty